Assay Details
| Gene Symbol: SLC4A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021196.3 | NP_067019.3 | 22 - 23 | 2962 | 72 | |
| GenBank mRNA | AF243499.2 | - | 22 - 23 | 2960 | 72 | |
More Information
Set Membership: |
|

SLC4A5
Genomic Map