Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052480.hg.1 |
TC01003356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052480.hg.1 |
ENST00000361409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052480.hg.1 |
ENST00000368194.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052480.hg.1 |
NM_014784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052480.hg.1 |
NM_198236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
ENST00000361409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
ENST00000368194.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
NM_014784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
NM_198236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
ENST00000361409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
ENST00000368194.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
NM_014784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
NM_198236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221712.hg.1 |
TC0100016027.hg.1 |
Clariom D TC |