Assay Details
| Gene Symbol: PROM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK026052.1 | - | 1 | 933 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium voltage-gated channel interacting protein 3 | KCNIP3 |
CSEN, DREAM, KCHIP3 |
Sense | Chr. 2 - 95297324 - 95386077 on NCBI Build GRCh38 | |
| Downstream | fumarylacetoacetate hydrolase domain containing 2A | FAHD2A | CGI-105 | 102.4kb | Sense | Chr. 2 - 95402691 - 95416702 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PROM2
Genomic Map