Assay Details
| Gene Symbol: SRSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036610.1 | 5 - 6 | 1005 | 110 | ||
| GenBank mRNA | AK310317.1 | - | 1583 | 110 | ||
| BM833102.1 | - | 3 - 4 | 561 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000530.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
mRNA 3'-end processing
regulation of mRNA splicing, via spliceosome
nucleotide binding
RNA binding
protein binding
phospholipase binding
poly(A) RNA binding
sequence-specific mRNA binding
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
mRNA 3'-end processing
regulation of mRNA splicing, via spliceosome
nucleotide binding
RNA binding
protein binding
phospholipase binding
poly(A) RNA binding
sequence-specific mRNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06005787.hg.1 | TC06000530.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005787.hg.1 | ENST00000373715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005787.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005787.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154409.hg.1 | TC0600007842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |

SRSF3
Genomic Map