Assay Details
Gene Symbol: SCIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112706.2 | NP_001106177.1 | 3 - 4 | 729 | 59 | |
GenBank mRNA | AB067492.1 | - | 3 - 4 | 729 | 59 | |
AF276507.1 | - | 3 - 4 | 519 | 59 | ||
AK075123.1 | - | 3 - 4 | 588 | 59 | ||
AK290363.1 | - | 3 - 4 | 622 | 59 | ||
BC021090.1 | - | 3 - 4 | 568 | 4877906 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
