Assay Details
Gene Symbol: FKSG29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024013.1 | 1 | 460 | 75 | ||
GenBank mRNA | AY014273.1 | - | 1 | 460 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | UBA domain containing 2 | UBAC2 | PHGDHL1 | Anti-sense | Chr. 13 - 99200425 - 99386499 on NCBI Build GRCh38 | |
Downstream | microRNA 623 | MIR623 |
MIRN623, hsa-mir-623 |
4.2kb | Anti-sense | Chr. 13 - 99356131 - 99356228 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000354.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P119813 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P119813 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 19222 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 54603 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P119813 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P119813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523951 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074737 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523973 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074765 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523991 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074768 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523992 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523970 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523965 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523981 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074748 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074764 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074737 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523991 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074768 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523965 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074765 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523970 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074764 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523992 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523951 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523981 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523973 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16663181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09074748 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29523941 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P119813 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P119813 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074780 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16663184 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074774 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16663182 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074769 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074776 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074778 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074764 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074744 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074736 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074761 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074771 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16663180 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074752 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074729 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074734 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074766 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074786 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074782 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074740 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074788 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074758 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074733 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074748 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074756 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074750 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074754 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074784 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16663181 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074737 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09074767 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 223896_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11757255_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11757255_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR13003742.hg.1 | NR_024013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13003742.hg.1 | TC13000354.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1300152743.hg.1 | NR_024013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300152743.hg.1 | NR_024013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300152743.hg.1 | TC1300007833.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 223896_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 223896_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 223896_at8 | - | - |
Human Genome U133 Set | 223896_PM_at | - | - |
Human Genome U133 Set | 223896_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050421 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050421 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 461230 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3238555 | NR_024013.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3238555 | AY014273.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3238555 | NR_024013.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3238555 | AY014273.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238555 | NR_024013.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238555 | AY014273.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238555 | NR_024013.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238555 | AY014273.1 | human_transcriptome |
