Assay Details
| Gene Symbol: TAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003185.3 | NP_003176.2 | 10 - 11 | 2658 | 98 | |
| GenBank mRNA | AK123937.1 | - | 6 - 7 | 1507 | 98 | |
| U75308.1 | - | 12 - 13 | 2244 | 98 | ||
| Y11354.1 | - | 10 - 11 | 2652 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001004.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
DNA-templated transcription, initiation
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
viral process
positive regulation of transcription, DNA-templated
regulation of signal transduction by p53 class mediator
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
protein heterodimerization activity
ovarian follicle development
DNA-templated transcription, initiation
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
viral process
positive regulation of transcription, DNA-templated
regulation of signal transduction by p53 class mediator
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20013670.hg.1 | ENST00000252996.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013670.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013670.hg.1 | TC20001004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000486599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000486599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000486599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000608887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000608887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000608887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000486599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000486599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000486599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000488539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000608458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000608887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000608887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000608887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | ENST00000609045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010031.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171933.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171933.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171933.hg.1 | NM_003185.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171933.hg.1 | TC2000010031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171933.hg.1 | ENST00000252996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171933.hg.1 | ENST00000436129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171933.hg.1 | NM_003185.3 | Refseq |

TAF4
Genomic Map