Assay Details
| Gene Symbol: PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015545.3 | NP_056360.2 | 3 - 4 | 762 | 64 | |
| GenBank mRNA | AB014532.1 | - | 3 - 4 | 670 | 64 | |
| AK001615.1 | - | 3 - 4 | 677 | 64 | ||
| AK291004.1 | - | 3 - 4 | 653 | 64 | ||
| BC003688.1 | - | 2 - 3 | 275 | 64 | ||
| BC080580.1 | - | 3 - 4 | 710 | 6645909 | 64 | |
| BC103495.1 | - | 3 - 4 | 687 | 40011658 | 64 | |
| BC103502.1 | - | 3 - 4 | 687 | 40011661 | 64 | |
| Gene Symbol: ATP5J2-PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198879.1 | NP_001185808.1 | 4 - 5 | 795 | 64 | |
| GenBank mRNA | AK295912.1 | - | 4 - 5 | 758 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003365.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025060.hg.1 | TC07003365.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025060.hg.1 | ENST00000292478.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025060.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025060.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201289.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201289.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201289.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201289.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201289.hg.1 | TC0700013592.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201238.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201238.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201238.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201238.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201238.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201238.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201238.hg.1 | TC0700013590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |

PTCD1
Genomic Map