Assay Details
| Gene Symbol: PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015545.3 | NP_056360.2 | 1 - 2 | 146 | 115 | |
| GenBank mRNA | BC080580.1 | - | 1 - 2 | 94 | 6645909 | 115 |
| BG717613.1 | - | 1 - 2 | 145 | 115 | ||
| DA770648.1 | - | 1 - 2 | 146 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003365.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC080580.1,NM_015545.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07025077.hg.1 | TC07003365.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025077.hg.1 | ENST00000292478.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025077.hg.1 | ENST00000438524.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025077.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025072.hg.1 | TC07003365.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025072.hg.1 | ENST00000292478.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025072.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025071.hg.1 | TC07003365.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025071.hg.1 | ENST00000292478.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025071.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025081.hg.1 | TC07003365.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025081.hg.1 | ENST00000292478.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025081.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201252.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201252.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201252.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201252.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201252.hg.1 | TC0700013590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201244.hg.1 | TC0700013590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | ENST00000485746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013590.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | ENST00000430029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201245.hg.1 | TC0700013590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700201249.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201249.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201249.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201249.hg.1 | ENST00000292478.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201249.hg.1 | ENST00000419981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201249.hg.1 | NM_015545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201249.hg.1 | TC0700013590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000413834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000430982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000437572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | ENST00000451138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013592.hg.1 | NM_001198879.1 | Refseq |

PTCD1
Genomic Map