Assay Details
Gene Symbol: UNC93A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011535905.2 | XP_011534207.1 | 1 - 2 | 534 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011535905.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000230256.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000366830.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000503433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | ENST00000504706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_001143947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010210.hg.1 | NM_018974.3 | Refseq |
