Assay Details
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160042.1 | NP_001153514.1 | 2 - 3 | 474 | 63 | |
| NM_018134.2 | NP_060604.2 | 2 - 3 | 234 | 63 | ||
| GenBank mRNA | AK001409.1 | - | 2 - 3 | 198 | 63 | |
| AK223272.1 | - | 2 - 3 | 222 | 63 | ||
| AK307853.1 | - | 2 - 3 | 438 | 63 | ||
| BC098250.1 | - | 2 - 3 | 197 | 63 | ||
| BC098260.1 | - | 2 - 3 | 197 | 40024457 | 63 | |
| BC098317.1 | - | 2 - 3 | 197 | 40024460 | 63 | |
| BC098366.1 | - | 2 - 3 | 197 | 40024462 | 63 | |
| DA746781.1 | - | 2 - 3 | 234 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000422.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01006461.hg.1 | TC01000422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006461.hg.1 | ENST00000291358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006461.hg.1 | ENST00000537469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006461.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006461.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006460.hg.1 | TC01000422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006460.hg.1 | ENST00000537469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006460.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006459.hg.1 | TC01000422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006459.hg.1 | ENST00000291358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006459.hg.1 | ENST00000537469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006459.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006459.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006457.hg.1 | TC01000422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006457.hg.1 | ENST00000537469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006457.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006457.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006463.hg.1 | TC01000422.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006463.hg.1 | ENST00000291358.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006463.hg.1 | ENST00000537469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006463.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006463.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153866.hg.1 | TC0100007670.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007670.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153862.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153862.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153862.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153862.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153862.hg.1 | TC0100007670.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100153867.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153867.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153867.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153867.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153867.hg.1 | TC0100007670.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153870.hg.1 | TC0100007670.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | ENST00000291358.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | ENST00000537469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | ENST00000617816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | NM_001160042.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | NM_018134.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153868.hg.1 | TC0100007670.hg.1 | Clariom D TC |

IQCC
Genomic Map