Assay Details
| Gene Symbol: ZSWIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020883.1 | NP_065934.1 | 4 - 5 | 1475 | 103 | |
| XM_011541861.2 | XP_011540163.1 | 4 - 5 | 1483 | 103 | ||
| XM_017001913.1 | XP_016857402.1 | 3 - 4 | 561 | 103 | ||
| GenBank mRNA | AB040944.1 | - | 4 - 5 | 1453 | 103 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002605.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01040517.hg.1 | TC01002605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040517.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040517.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040518.hg.1 | TC01002605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040518.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040518.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009874.hg.1 | ENST00000420606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009874.hg.1 | ENST00000420606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010060.hg.1 | ENST00000420212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010060.hg.1 | ENST00000420212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010060.hg.1 | ENST00000420212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040515.hg.1 | TC01002605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040515.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040515.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040514.hg.1 | TC01002605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040514.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040514.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040512.hg.1 | TC01002605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040512.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040512.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040516.hg.1 | TC01002605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040516.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040516.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000464588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000464588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | ENST00000464588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040513.hg.1 | TC01002605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040513.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040513.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204634.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204634.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204634.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204634.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204634.hg.1 | TC0100013999.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100204631.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204631.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204631.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204631.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204631.hg.1 | TC0100013999.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100204636.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204636.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204636.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204636.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204636.hg.1 | TC0100013999.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010060.hg.1 | ENST00000420212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010060.hg.1 | ENST00000420212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010060.hg.1 | ENST00000420212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204635.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204635.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204635.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204635.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204635.hg.1 | TC0100013999.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100204632.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204632.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204632.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204632.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204632.hg.1 | TC0100013999.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100204633.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204633.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204633.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204633.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204633.hg.1 | TC0100013999.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009874.hg.1 | ENST00000420606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009874.hg.1 | ENST00000420606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204630.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204630.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204630.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204630.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204630.hg.1 | TC0100013999.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000359600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000464588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000464588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | ENST00000464588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013999.hg.1 | NM_020883.1 | Refseq |

ZSWIM5
Genomic Map