Assay Details
| Gene Symbol: SPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030572.3 | NP_085049.1 | 1 - 2 | 219 | 110 | |
| GenBank mRNA | AK075342.1 | - | 1 - 2 | 173 | 110 | |
| BC004336.1 | - | 1 - 2 | 154 | 3631700 | 110 | |
| DA380664.1 | - | 1 - 2 | 219 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000234.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK075342.1,BC004336.1,NM_030572.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12002904.hg.1 | ENST00000256969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002904.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002904.hg.1 | TC12000234.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002905.hg.1 | ENST00000256969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002905.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002905.hg.1 | TC12000234.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002906.hg.1 | ENST00000256969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002906.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002906.hg.1 | TC12000234.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200149486.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149486.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149486.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149486.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149486.hg.1 | TC1200007053.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200149484.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149484.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149484.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149484.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149484.hg.1 | TC1200007053.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000256969.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535033.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000535139.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000537527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000543800.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000544637.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | ENST00000546199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007053.hg.1 | NM_030572.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 195982 | - | - |

SPX
Genomic Map