Assay Details
| Gene Symbol: M6PR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002355.3 | NP_002346.1 | 4 - 5 | 731 | 63 | |
| XM_005253376.2 | XP_005253433.1 | 4 - 5 | 593 | 63 | ||
| GenBank mRNA | AK222792.1 | - | 4 - 5 | 612 | 63 | |
| AK291143.1 | - | 4 - 5 | 577 | 63 | ||
| BC017111.1 | - | 2 - 3 | 131 | 63 | ||
| BC024206.1 | - | 4 - 5 | 623 | 3530887 | 63 | |
| CR457412.1 | - | 3 - 4 | 456 | 63 | ||
| M16985.1 | - | 4 - 5 | 601 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001184.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000000412.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000536844.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000537621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000539143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000540837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000541507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000543845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | ENST00000544245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_001207024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009843.hg.1 | NM_002355.3 | Refseq |

M6PR
Genomic Map