Assay Details
| Gene Symbol: DNAH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369.2 | NP_001360.1 | 54 - 55 | 9146 | 151 | |
| XM_005248262.3 | XP_005248319.2 | 54 - 55 | 9229 | 151 | ||
| XM_017009177.1 | XP_016864666.1 | 54 - 55 | 9229 | 151 | ||
| XM_017009178.1 | XP_016864667.1 | 54 - 55 | 10032 | 151 | ||
| XM_017009179.1 | XP_016864668.1 | 48 - 49 | 8326 | 151 | ||
| XM_017009180.1 | XP_016864669.1 | 54 - 55 | 9229 | 151 | ||
| XM_017009181.1 | XP_016864670.1 | 54 - 55 | 9229 | 151 | ||
| XM_017009182.1 | XP_016864671.1 | 54 - 55 | 9229 | 151 | ||
| XM_017009183.1 | XP_016864672.1 | 54 - 55 | 9229 | 151 | ||
| XM_017009185.1 | XP_016864674.1 | 26 - 27 | 4525 | 151 | ||
| XM_017009186.1 | XP_016864675.1 | 23 - 24 | 3941 | 151 | ||
| XM_017009188.1 | XP_016864677.1 | 20 - 21 | 4757 | 151 | ||
| GenBank mRNA | AB046823.2 | - | 2 - 3 | 210 | 151 | |
| AB290164.1 | - | 7 - 8 | 1149 | 151 | ||
| AY045575.1 | - | 54 - 55 | 9146 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001171.hg.1
Set Membership: |
|

DNAH5
Genomic Map