Assay Details
| Gene Symbol: LOC101928441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL517295.3 | - | 1 - 2 | 108 | 85 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000534841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000534841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000534841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000534841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000534841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000534841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000541288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | ENST00000545400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | NR_120471.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007076.hg.1 | NR_120471.1 | Refseq |

LOC101928441
Genomic Map