Assay Details
| Gene Symbol: SLC45A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012509.3 | NP_001012527.1 | 5 - 6 | 1249 | 103 | |
| NM_016180.4 | NP_057264.3 | 5 - 6 | 1249 | 103 | ||
| GenBank mRNA | AF172849.1 | - | 5 - 6 | 1182 | 103 | |
| BC064405.1 | - | 5 - 6 | 1215 | 5563039 | 103 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001245.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05017477.hg.1 | TC05001245.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017477.hg.1 | ENST00000382102.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017477.hg.1 | NM_001012509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017478.hg.1 | TC05001245.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017478.hg.1 | ENST00000296589.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017478.hg.1 | ENST00000342059.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017478.hg.1 | ENST00000382102.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017478.hg.1 | NM_001012509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017478.hg.1 | NM_016180.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017479.hg.1 | TC05001245.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017479.hg.1 | ENST00000296589.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017479.hg.1 | ENST00000342059.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017479.hg.1 | ENST00000382102.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017479.hg.1 | NM_001012509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017479.hg.1 | NM_016180.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172658.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172658.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172658.hg.1 | TC0500010456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500172661.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172661.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172661.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172661.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172661.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172661.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172661.hg.1 | TC0500010456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500172660.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172660.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172660.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172660.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172660.hg.1 | TC0500010456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000296589.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000382102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000505056.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000509381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | ENST00000510600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010456.hg.1 | NM_001297417.2 | Refseq |

SLC45A2
Genomic Map