Assay Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129758.1 | NP_001123230.1 | 1 - 2 | 459 | 81 | |
| NM_022357.3 | NP_071752.3 | 1 - 2 | 459 | 81 | ||
| GenBank mRNA | AJ291679.1 | - | 1 - 2 | 373 | 81 | |
| AK057401.1 | - | 1 - 2 | 459 | 81 | ||
| AY358390.1 | - | 1 - 2 | 382 | 81 | ||
| BC037243.1 | - | 1 - 2 | 371 | 81 | ||
| BC057789.1 | - | 1 - 2 | 400 | 5297958 | 81 | |
| DB020973.1 | - | 1 - 2 | 422 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001200.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016767.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016767.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016767.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016767.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016768.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016768.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016768.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016768.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016770.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016770.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016770.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016770.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016771.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016771.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016771.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016771.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182489.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182492.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182492.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182492.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182492.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182492.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182492.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182492.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182488.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182488.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182488.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182488.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182488.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182488.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182488.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182490.hg.1 | TC1600010664.hg.1 | Clariom D TC |

DPEP3
Genomic Map