| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016766.hg.1 |
ENST00000268793.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016766.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016766.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16016766.hg.1 |
TC16001200.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182487.hg.1 |
TC1600010664.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
ENST00000574342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010664.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182486.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600182486.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182486.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182486.hg.1 |
ENST00000268793.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600182486.hg.1 |
NM_001129758.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182486.hg.1 |
NM_022357.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600182486.hg.1 |
TC1600010664.hg.1 |
Clariom D TC |