Assay Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129758.1 | NP_001123230.1 | 6 - 7 | 86 | ||
| NM_022357.3 | NP_071752.3 | 6 - 7 | 1098 | 89 | ||
| GenBank mRNA | AJ291679.1 | - | 6 - 7 | 1012 | 89 | |
| AK057401.1 | - | 6 - 7 | 1098 | 89 | ||
| AY358390.1 | - | 6 - 7 | 86 | |||
| BC037243.1 | - | 6 - 7 | 1010 | 89 | ||
| BC057789.1 | - | 6 - 7 | 1039 | 5297958 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001200.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016759.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016759.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016759.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016759.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016758.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016758.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016758.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016758.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016760.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016760.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016760.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016760.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016761.hg.1 | ENST00000268793.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016761.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016761.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016761.hg.1 | TC16001200.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600182477.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182477.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182477.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182477.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182477.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182477.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182477.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182479.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182479.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182479.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182479.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182479.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182479.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182479.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182481.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182481.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182481.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182481.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182481.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182481.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182481.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182478.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182478.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182478.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182478.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182478.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182478.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182478.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182480.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182480.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182480.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182480.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182480.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182480.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182480.hg.1 | TC1600010664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000268793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | ENST00000574342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_001129758.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010664.hg.1 | NM_022357.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 154469 | - | - |

DPEP3
Genomic Map