Assay Details
Gene Symbol: FAM57A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318006.1 | NP_001304935.1 | 2 - 3 | 309 | 155 | |
NM_024792.2 | NP_079068.1 | 2 - 3 | 309 | 155 | ||
GenBank mRNA | AF477201.1 | - | 2 - 3 | 297 | 155 | |
AK025935.1 | - | 2 - 3 | 248 | 155 | ||
AK292065.1 | - | 2 - 3 | 248 | 155 | ||
BC026023.1 | - | 2 - 3 | 271 | 4548729 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000301324.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000308278.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000570892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000572018.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000574327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | ENST00000577008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006464.hg.1 | NM_024792.2 | Refseq |
