Assay Details
| Gene Symbol: IL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199640.1 | NP_001186569.1 | 1 - 2 | 60 | 118 | |
| NM_001199641.1 | NP_001186570.1 | 1 - 2 | 60 | 118 | ||
| NM_001314046.1 | NP_001300975.1 | 1 - 2 | 60 | 118 | ||
| NM_001314048.1 | NP_001300977.1 | 1 - 2 | 60 | 118 | ||
| NM_033439.3 | NP_254274.1 | 1 - 2 | 60 | 118 | ||
| GenBank mRNA | DB199599.1 | - | 1 - 2 | 69 | 118 | |
| DB477860.1 | - | 1 - 2 | 60 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000045.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001199640.1,NM_001199641.1,NM_001314046.1,NM_001314048.1,NM_033439.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of leukocyte migration
negative regulation of T-helper 1 type immune response
positive regulation of type 2 immune response
transcription, DNA-templated
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of interferon-gamma production
positive regulation of interleukin-13 production
positive regulation of interleukin-4 production
positive regulation of interleukin-5 production
positive regulation of interleukin-6 production
type 2 immune response
positive regulation of macrophage activation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of inflammatory response
positive regulation of immunoglobulin secretion
negative regulation of immunoglobulin secretion
defense response to virus
positive regulation of chemokine secretion
extrinsic apoptotic signaling pathway
cytokine activity
protein binding
negative regulation of leukocyte migration
negative regulation of T-helper 1 type immune response
positive regulation of type 2 immune response
transcription, DNA-templated
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of interferon-gamma production
positive regulation of interleukin-13 production
positive regulation of interleukin-4 production
positive regulation of interleukin-5 production
positive regulation of interleukin-6 production
type 2 immune response
positive regulation of macrophage activation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of inflammatory response
positive regulation of immunoglobulin secretion
negative regulation of immunoglobulin secretion
defense response to virus
positive regulation of chemokine secretion
extrinsic apoptotic signaling pathway
cytokine activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | TC09000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | ENST00000381434.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | ENST00000417746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | ENST00000456383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | ENST00000463336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000588.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000463336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000463336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000585.hg.1 | TC09000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000585.hg.1 | ENST00000417746.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000585.hg.1 | ENST00000456383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000585.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000585.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000585.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145018.hg.1 | TC0900006577.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000463336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000463336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006577.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000463336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000381434.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000417746.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000456383.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000463336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | ENST00000611532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | NM_001199640.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | NM_001199641.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | NM_033439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145023.hg.1 | TC0900006577.hg.1 | Clariom D TC |

IL33
Genomic Map