Assay Details
| Gene Symbol: UTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007124.2 | NP_009055.2 | 38 - 39 | 5636 | 158 | |
| XM_005267127.4 | XP_005267184.1 | 38 - 39 | 5971 | 158 | ||
| XM_005267130.2 | XP_005267187.1 | 39 - 40 | 5854 | 158 | ||
| XM_005267133.2 | XP_005267190.1 | 38 - 39 | 6654 | 158 | ||
| XM_011536101.2 | XP_011534403.1 | 40 - 41 | 6315 | 158 | ||
| XM_011536102.2 | XP_011534404.1 | 40 - 41 | 5856 | 158 | ||
| XM_011536106.2 | XP_011534408.1 | 39 - 40 | 6503 | 158 | ||
| XM_011536107.2 | XP_011534409.1 | 39 - 40 | 5879 | 158 | ||
| XM_017011243.1 | XP_016866732.1 | 40 - 41 | 5853 | 158 | ||
| XM_017011244.1 | XP_016866733.1 | 40 - 41 | 5856 | 158 | ||
| XM_017011245.1 | XP_016866734.1 | 40 - 41 | 5856 | 158 | ||
| GenBank mRNA | X69086.1 | - | 38 - 39 | 5544 | 158 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001059.hg.1
Set Membership: |
|

UTRN
Genomic Map