Assay Details
| Gene Symbol: UTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007124.2 | NP_009055.2 | 41 - 42 | 6146 | 69 | |
| XM_005267127.4 | XP_005267184.1 | 41 - 42 | 6481 | 69 | ||
| XM_005267130.2 | XP_005267187.1 | 42 - 43 | 6364 | 69 | ||
| XM_005267133.2 | XP_005267190.1 | 41 - 42 | 7164 | 69 | ||
| XM_011536101.2 | XP_011534403.1 | 43 - 44 | 6825 | 69 | ||
| XM_011536102.2 | XP_011534404.1 | 43 - 44 | 6366 | 69 | ||
| XM_011536106.2 | XP_011534408.1 | 42 - 43 | 7013 | 69 | ||
| XM_011536107.2 | XP_011534409.1 | 42 - 43 | 6389 | 69 | ||
| XM_017011243.1 | XP_016866732.1 | 43 - 44 | 6363 | 69 | ||
| XM_017011244.1 | XP_016866733.1 | 43 - 44 | 6366 | 69 | ||
| XM_017011245.1 | XP_016866734.1 | 43 - 44 | 6366 | 69 | ||
| GenBank mRNA | X69086.1 | - | 41 - 42 | 6054 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001059.hg.1
Set Membership: |
|

UTRN
Genomic Map