Assay Details
| Gene Symbol: UTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007124.2 | NP_009055.2 | 49 - 50 | 7387 | 115 | |
| XM_005267127.4 | XP_005267184.1 | 49 - 50 | 7722 | 115 | ||
| XM_005267130.2 | XP_005267187.1 | 50 - 51 | 7605 | 115 | ||
| XM_005267133.2 | XP_005267190.1 | 49 - 50 | 8405 | 115 | ||
| XM_011536101.2 | XP_011534403.1 | 51 - 52 | 8066 | 115 | ||
| XM_011536102.2 | XP_011534404.1 | 51 - 52 | 7607 | 115 | ||
| XM_011536106.2 | XP_011534408.1 | 50 - 51 | 8254 | 115 | ||
| XM_011536107.2 | XP_011534409.1 | 50 - 51 | 7630 | 115 | ||
| XM_017011243.1 | XP_016866732.1 | 51 - 52 | 7604 | 115 | ||
| XM_017011244.1 | XP_016866733.1 | 51 - 52 | 7607 | 115 | ||
| XM_017011245.1 | XP_016866734.1 | 51 - 52 | 7607 | 115 | ||
| GenBank mRNA | X69086.1 | - | 49 - 50 | 7295 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001059.hg.1
Set Membership: |
|

UTRN
Genomic Map