| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06012492.hg.1 |
TC06001059.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06012492.hg.1 |
ENST00000367526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06012492.hg.1 |
ENST00000367545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06012492.hg.1 |
NM_007124.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600166625.hg.1 |
ENST00000367526.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600166625.hg.1 |
ENST00000367545.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600166625.hg.1 |
NM_007124.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600166625.hg.1 |
ENST00000367526.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600166625.hg.1 |
ENST00000367545.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600166625.hg.1 |
NM_007124.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600166625.hg.1 |
TC0600009712.hg.1 |
Clariom D TC |