Assay Details
| Gene Symbol: IL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314045.1 | NP_001300974.1 | 1 - 2 | 54 | 105 | |
| XM_017015285.1 | XP_016870774.1 | 3 - 4 | 805 | 105 | ||
| GenBank mRNA | DA439244.1 | - | 1 - 2 | 54 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001314045.1, XM_017015285.1 transcripts.
Set Membership: |
|

IL33
Genomic Map