| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025593.hg.1 |
TC05001864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025593.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025593.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025595.hg.1 |
TC05001864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025595.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025595.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025596.hg.1 |
TC05001864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025596.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025596.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025594.hg.1 |
TC05001864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025594.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025594.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197002.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197002.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197002.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197002.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197002.hg.1 |
TC0500013376.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500197006.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197006.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197006.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197006.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197006.hg.1 |
TC0500013376.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500197004.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197004.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197004.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197004.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197004.hg.1 |
TC0500013376.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013376.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197003.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197003.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197003.hg.1 |
ENST00000542735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500197003.hg.1 |
NM_194249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500197003.hg.1 |
TC0500013376.hg.1 |
Clariom D TC |