Assay Details
| Gene Symbol: PRSS30P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC041609.1 | - | 1 - 2 | 165 | 64 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000390681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000475030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000476276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000480827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000489864.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | ENST00000506153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009146.hg.1 | NR_026864.2 | Refseq |

PRSS30P
Genomic Map