Assay Details
| Gene Symbol: CD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040033.1 | NP_001035122.1 | 1 - 2 | 65 | 60 | |
| GenBank mRNA | BC035456.1 | - | 1 - 2 | 47 | 60 | |
| CA453414.1 | - | 67 | 60 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000968.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001040033.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015420.hg.1 | TC01000968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015420.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015420.hg.1 | ENST00000429072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015420.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015420.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015413.hg.1 | TC01000968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015413.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167539.hg.1 | TC0100009394.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000271324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000464329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000471220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000476408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | ENST00000497404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_000560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009394.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167534.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167534.hg.1 | NM_001040033.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167534.hg.1 | TC0100009394.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 159670 | - | - |

CD53
Genomic Map