Assay Details
| Gene Symbol: SQRDL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271213.1 | NP_001258142.1 | 8 - 9 | 1244 | 137 | |
| NM_021199.3 | NP_067022.1 | 7 - 8 | 1437 | 137 | ||
| GenBank mRNA | AF042284.1 | - | 7 - 8 | 1127 | 137 | |
| AF118085.1 | - | 6 - 7 | 958 | 137 | ||
| AF151802.1 | - | 7 - 8 | 1121 | 137 | ||
| AK130140.1 | - | 8 - 9 | 1244 | 137 | ||
| AK222463.1 | - | 7 - 8 | 1136 | 137 | ||
| AK313111.1 | - | 7 - 8 | 1116 | 137 | ||
| BC016836.1 | - | 7 - 8 | 1092 | 3884024 | 137 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000364.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15003333.hg.1 | ENST00000260324.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003333.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003333.hg.1 | TC15000364.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182330.hg.1 | TC1500010737.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | ENST00000260324.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | ENST00000565997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | ENST00000568606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | NM_001271213.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | NM_021199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500182329.hg.1 | TC1500010737.hg.1 | Clariom D TC |

SQRDL
Genomic Map