Assay Details
| Gene Symbol: TIPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017858.2 | NP_060328.2 | 4 - 5 | 379 | 93 | |
| XM_005254521.2 | XP_005254578.1 | 4 - 5 | 389 | 93 | ||
| XM_005254522.4 | XP_005254579.1 | 4 - 5 | 366 | 93 | ||
| XM_006720594.3 | XP_006720657.1 | 4 - 5 | 431 | 93 | ||
| XM_011521743.1 | XP_011520045.1 | 4 - 5 | 562 | 93 | ||
| XM_017022390.1 | XP_016877879.1 | 5 - 6 | 445 | 93 | ||
| GenBank mRNA | AK000523.1 | - | 4 - 5 | 317 | 93 | |
| BC000870.1 | - | 4 - 5 | 354 | 3460338 | 93 | |
| BU596053.1 | - | 4 - 5 | 381 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001576.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication checkpoint
DNA replication
mitotic nuclear division
positive regulation of cell proliferation
response to UV
intra-S DNA damage checkpoint
regulation of nuclear cell cycle DNA replication
cell cycle phase transition
replication fork protection
cell division
protein binding
DNA replication checkpoint
DNA replication
mitotic nuclear division
positive regulation of cell proliferation
response to UV
intra-S DNA damage checkpoint
regulation of nuclear cell cycle DNA replication
cell cycle phase transition
replication fork protection
cell division
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15015174.hg.1 | ENST00000261881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015174.hg.1 | ENST00000367709.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015174.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015174.hg.1 | TC15001576.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015172.hg.1 | ENST00000261881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015172.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015172.hg.1 | TC15001576.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015175.hg.1 | ENST00000261881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015175.hg.1 | ENST00000367709.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015175.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015175.hg.1 | TC15001576.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015176.hg.1 | ENST00000261881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015176.hg.1 | ENST00000367709.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015176.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015176.hg.1 | TC15001576.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009764.hg.1 | ENST00000425348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015171.hg.1 | ENST00000261881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015171.hg.1 | ENST00000367709.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015171.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015171.hg.1 | TC15001576.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010881.hg.1 | ENST00000489563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184967.hg.1 | TC1500010880.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010881.hg.1 | ENST00000489563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010881.hg.1 | ENST00000561773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184965.hg.1 | TC1500010880.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184966.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184966.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184966.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184966.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184966.hg.1 | TC1500010880.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009764.hg.1 | ENST00000425348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010880.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184970.hg.1 | TC1500010880.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184972.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184972.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184972.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184972.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184972.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184972.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184972.hg.1 | TC1500010880.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | ENST00000261881.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | ENST00000566524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | NM_001289986.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | NM_017858.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184968.hg.1 | TC1500010880.hg.1 | Clariom D TC |

TIPIN
Genomic Map