Assay Details
| Gene Symbol: KLRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114396.1 | NP_001107868.1 | 5 - 6 | 511 | 89 | |
| NM_002262.3 | NP_002253.1 | 4 - 5 | 574 | 89 | ||
| NM_007334.2 | NP_031360.1 | 3 - 4 | 481 | 89 | ||
| XM_017019287.1 | XP_016874776.1 | 5 - 6 | 516 | 89 | ||
| XM_017019289.1 | XP_016874778.1 | 3 - 4 | 478 | 89 | ||
| XM_017019291.1 | XP_016874780.1 | 5 - 6 | 498 | 89 | ||
| GenBank mRNA | AB009597.1 | - | 3 - 4 | 258 | 89 | |
| AK291506.1 | - | 5 - 6 | 516 | 89 | ||
| AY227806.1 | - | 5 - 6 | 511 | 89 | ||
| AY327501.1 | - | 3 - 4 | 379 | 89 | ||
| BC028009.1 | - | 4 - 5 | 387 | 5213081 | 89 | |
| U30610.1 | - | 4 - 5 | 398 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000160.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
natural killer cell mediated immunity
cell surface receptor signaling pathway
innate immune response
regulation of immune response
transmembrane signaling receptor activity
protein binding
MHC class I protein complex binding
MHC class Ib protein binding, via antigen binding groove
carbohydrate binding
protein antigen binding
natural killer cell mediated immunity
cell surface receptor signaling pathway
innate immune response
regulation of immune response
transmembrane signaling receptor activity
protein binding
MHC class I protein complex binding
MHC class Ib protein binding, via antigen binding groove
carbohydrate binding
protein antigen binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000336164.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000344825.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000350274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000381908.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000538997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000538997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000538997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000538997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000538997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000538997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000538997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000539792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000540271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000543777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | ENST00000544747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_001114396.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_002262.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006788.hg.1 | NM_007334.2 | Refseq |

KLRD1
Genomic Map