Assay Details
| Gene Symbol: SLC24A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005251426.4 | XP_005251483.1 | 1 - 2 | 807 | 139 | |
| XM_006716750.3 | XP_006716813.1 | 1 - 2 | 807 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005251426.4, XM_006716750.3 transcripts.
Set Membership: |
|

SLC24A2
Genomic Map