Assay Details
| Gene Symbol: VCPKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040662.1 | NP_001035752.1 | 4 - 5 | 599 | 164 | |
| GenBank mRNA | BC143674.1 | - | 4 - 5 | 620 | 9052188 | 164 |
| BI767189.1 | - | 4 - 5 | 570 | 164 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001115.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14012536.hg.1 | ENST00000358490.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012536.hg.1 | ENST00000395859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012536.hg.1 | ENST00000395860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012536.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012536.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012536.hg.1 | TC14001115.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009122.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000395859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000395860.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000484763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000491402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | ENST00000569518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NM_001040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NM_024558.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NR_049738.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | NR_049739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167209.hg.1 | TC1400009122.hg.1 | Clariom D TC |

VCPKMT
Genomic Map