Assay Details
| Gene Symbol: ATL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290048.1 | NP_001276977.1 | 10 - 11 | 1325 | 59 | |
| NM_015459.4 | NP_056274.3 | 10 - 11 | 1427 | 59 | ||
| XM_006718493.1 | XP_006718556.1 | 9 - 10 | 1252 | 59 | ||
| GenBank mRNA | AK023383.1 | - | 10 - 11 | 1032 | 59 | |
| AK097588.1 | - | 12 - 13 | 1367 | 59 | ||
| AK301910.1 | - | 10 - 11 | 1263 | 59 | ||
| BC077727.1 | - | 10 - 11 | 1318 | 5952471 | 59 | |
| CR936784.1 | - | 10 - 11 | 1310 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001894.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11022329.hg.1 | ENST00000332645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022329.hg.1 | ENST00000398868.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022329.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022329.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022329.hg.1 | TC11001894.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184842.hg.1 | TC1100011128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184841.hg.1 | TC1100011128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |

ATL3
Genomic Map