Assay Details
| Gene Symbol: ATL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290048.1 | NP_001276977.1 | 6 - 7 | 905 | 107 | |
| NM_015459.4 | NP_056274.3 | 6 - 7 | 1007 | 107 | ||
| GenBank mRNA | AK023383.1 | - | 6 - 7 | 612 | 107 | |
| AK097588.1 | - | 8 - 9 | 947 | 107 | ||
| AK301910.1 | - | 6 - 7 | 843 | 107 | ||
| AL117600.1 | - | 6 - 7 | 107 | |||
| BC077727.1 | - | 6 - 7 | 898 | 5952471 | 107 | |
| CR936784.1 | - | 6 - 7 | 890 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001894.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11022334.hg.1 | ENST00000332645.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022334.hg.1 | ENST00000398868.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022334.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022334.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022334.hg.1 | TC11001894.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184845.hg.1 | TC1100011128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |

ATL3
Genomic Map