Assay Details
| Gene Symbol: ATL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290048.1 | NP_001276977.1 | 1 - 2 | 338 | 62 | |
| GenBank mRNA | AK301910.1 | - | 1 - 2 | 276 | 62 | |
| DC394729.1 | - | 1 - 2 | 339 | 62 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK301910.1,NM_001290048.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184861.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184861.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184861.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184861.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184861.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184861.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184861.hg.1 | TC1100011128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100184863.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184863.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184863.hg.1 | TC1100011128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100184859.hg.1 | TC1100011128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000398868.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000535789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000538786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | ENST00000540699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_001290048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011128.hg.1 | NM_015459.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 116725 | - | - |

ATL3
Genomic Map