Assay Details
| Gene Symbol: LINC00282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027047.1 | 5 | 2250 | 83 | ||
| NR_027048.1 | 3 | 2107 | 83 | |||
| GenBank mRNA | AK094626.1 | - | 1 | 796 | 83 | |
| AK124707.1 | - | 3 | 2107 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dehydrogenase/reductase 12 | DHRS12 | SDR40C1 | -9.1kb | Sense | Chr. 13 - 51754856 - 51804206 on NCBI Build GRCh38 |
| Downstream | coiled-coil domain containing 70 | CCDC70 | 44.9kb | Anti-sense | Chr. 13 - 51861981 - 51866236 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC00282
Genomic Map