Assay Details
| Gene Symbol: ROCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005406.2 | NP_005397.1 | 10 - 11 | 2147 | 112 | |
| GenBank mRNA | AB208965.1 | - | 2 - 3 | 240 | 112 | |
| AK057038.1 | - | 10 - 11 | 2148 | 112 | ||
| AK307388.1 | - | 10 - 11 | 1962 | 112 | ||
| BC113114.1 | - | 10 - 11 | 1222 | 7961989 | 112 | |
| U43195.1 | - | 10 - 11 | 1206 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000397.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apical constriction
protein phosphorylation
cellular component disassembly involved in execution phase of apoptosis
smooth muscle contraction
leukocyte cell-cell adhesion
signal transduction
I-kappaB kinase/NF-kappaB signaling
Rho protein signal transduction
negative regulation of angiogenesis
membrane to membrane docking
actin cytoskeleton organization
regulation of cell adhesion
cortical actin cytoskeleton organization
bleb assembly
negative regulation of protein binding
regulation of actin cytoskeleton organization
negative regulation of myosin-light-chain-phosphatase activity
negative regulation of neuron apoptotic process
regulation of keratinocyte differentiation
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
leukocyte migration
leukocyte tethering or rolling
myoblast migration
regulation of stress fiber assembly
regulation of focal adhesion assembly
positive regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
regulation of establishment of endothelial barrier
negative regulation of bicellular tight junction assembly
regulation of establishment of cell polarity
regulation of cell motility
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
GTP-Rho binding
metal ion binding
apical constriction
protein phosphorylation
cellular component disassembly involved in execution phase of apoptosis
smooth muscle contraction
leukocyte cell-cell adhesion
signal transduction
I-kappaB kinase/NF-kappaB signaling
Rho protein signal transduction
negative regulation of angiogenesis
membrane to membrane docking
actin cytoskeleton organization
regulation of cell adhesion
cortical actin cytoskeleton organization
bleb assembly
negative regulation of protein binding
regulation of actin cytoskeleton organization
negative regulation of myosin-light-chain-phosphatase activity
negative regulation of neuron apoptotic process
regulation of keratinocyte differentiation
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
leukocyte migration
leukocyte tethering or rolling
myoblast migration
regulation of stress fiber assembly
regulation of focal adhesion assembly
positive regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
regulation of establishment of endothelial barrier
negative regulation of bicellular tight junction assembly
regulation of establishment of cell polarity
regulation of cell motility
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
GTP-Rho binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18004085.hg.1 | ENST00000399799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004085.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004085.hg.1 | TC18000397.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000578051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000578051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000578051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000582445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000582445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000582445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000583556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000583556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000583556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000584687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000584687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000584687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156147.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156147.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800156147.hg.1 | TC1800008219.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800156147.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800156147.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000399799.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000578051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000578051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000578051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000582445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000582445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000582445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000583556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000583556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000583556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000584687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000584687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000584687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | ENST00000584875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008219.hg.1 | NM_005406.2 | Refseq |

ROCK1
Genomic Map