Assay Details
| Gene Symbol: DCTD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005262779.3 | XP_005262836.1 | 2 - 3 | 306 | 70 | |
| XM_006714115.3 | XP_006714178.1 | 2 - 3 | 249 | 70 | ||
| XM_011531675.2 | XP_011529977.1 | 2 - 3 | 667 | 70 | ||
| XM_017007816.1 | XP_016863305.1 | 2 - 3 | 282 | 70 | ||
| XM_017007817.1 | XP_016863306.1 | 2 - 3 | 249 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005262779.3, XM_006714115.3, XM_011531675.2, XM_017007816.1, XM_017007817.1 transcripts.
Set Membership: |
|

DCTD
Genomic Map