| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
ENST00000367351.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009819.hg.1 |
NM_025217.3 |
Refseq |