Assay Details
| Gene Symbol: GNPTG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023782.1 | XP_016879271.1 | 3 - 4 | 211 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNPTG| Gene Symbol: GNPTG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023782.1 | XP_016879271.1 | 3 - 4 | 211 | 56 | |
Set Membership: |
|