Assay Details
Gene Symbol: C16orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206967.2 | NP_996850.1 | 2 - 3 | 199 | 69 | |
GenBank mRNA | AB115764.1 | - | 2 - 3 | 185 | 69 | |
CD674248.1 | - | 2 - 3 | 199 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001319.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB115764.1,NM_206967.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018276.hg.1 | ENST00000284245.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018276.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018276.hg.1 | TC16001319.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600194695.hg.1 | TC1600011575.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000284245.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602719.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | ENST00000602914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011575.hg.1 | NM_206967.2 | Refseq |
