Assay Details
| Gene Symbol: SEC14L5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014692.1 | NP_055507.1 | 5 - 6 | 659 | 82 | |
| GenBank mRNA | AB007880.2 | - | 5 - 6 | 659 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000138.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002183.hg.1 | ENST00000251170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002183.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002183.hg.1 | TC16000138.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600189244.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600189244.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600189244.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600189244.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600189244.hg.1 | TC1600011346.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600189243.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600189243.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600189243.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600189243.hg.1 | ENST00000251170.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600189243.hg.1 | ENST00000587469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600189243.hg.1 | NM_014692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600189243.hg.1 | TC1600011346.hg.1 | Clariom D TC |

SEC14L5
Genomic Map