Assay Details
| Gene Symbol: CLEC4GP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002931.2 | 5 - 6 | 535 | 129 | ||
| GenBank mRNA | AK090689.1 | - | 5 - 6 | 535 | 129 | |
| AK090734.1 | - | 5 - 6 | 532 | 129 | ||
| AK123140.1 | - | 5 - 6 | 516 | 129 | ||
| BC047593.1 | - | 3 - 4 | 424 | 129 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C-type lectin domain family 4 member M | CLEC4M |
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN |
-18.7kb | Sense | Chr. 19 - 7763052 - 7770559 on NCBI Build GRCh38 |
| Downstream | ecotropic viral integration site 5 like | EVI5L | 40.6kb | Sense | Chr. 19 - 7830275 - 7864976 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000137.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002169.hg.1 | ENST00000512667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002169.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002169.hg.1 | TC19000137.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900199325.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199325.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199325.hg.1 | TC1900011659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199325.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199325.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000328853.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000598081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000599020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | ENST00000620128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_001244856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009492.hg.1 | NM_198492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011659.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199327.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199327.hg.1 | NR_002931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199327.hg.1 | TC1900011659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199327.hg.1 | ENST00000596555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199327.hg.1 | NR_002931.2 | Refseq |

CLEC4GP1
Genomic Map