Assay Details
| Gene Symbol: LOC101928565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125956.1 | 1 - 2 | 56 | 81 | ||
| GenBank mRNA | AF085892.1 | - | 1 - 2 | 56 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224228.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224228.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224228.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224228.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224228.hg.1 | TC0100016335.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100224231.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224231.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224231.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224231.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224231.hg.1 | TC0100016335.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100224232.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224232.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224232.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224232.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224232.hg.1 | TC0100016335.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100224227.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224227.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224227.hg.1 | TC0100016335.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016335.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224229.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224229.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224229.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224229.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224229.hg.1 | TC0100016335.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100224230.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224230.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224230.hg.1 | ENST00000422253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224230.hg.1 | NR_125956.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224230.hg.1 | TC0100016335.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |

LOC101928565
Genomic Map