Assay Details
| Gene Symbol: LOC105372953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC023608.2 | - | 1 - 2 | 114 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929771 | LOC101929771 | -545.0kb | Sense | Chr. 1 - 221827666 - 221840666 on NCBI Build GRCh38 | |
| Downstream | HHIP like 2 | HHIPL2 | KIAA1822L | 134.8kb | Sense | Chr. 1 - 222501521 - 222548102 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105372953
Genomic Map