Assay Details
| Gene Symbol: LOC105372953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC023608.2 | - | 3 - 4 | 283 | 177 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929771 | LOC101929771 | -248.5kb | Sense | Chr. 1 - 221827666 - 221840666 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105372953
Genomic Map