Assay Details
| Gene Symbol: VAC14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034083.2 | 1 - 2 | 196 | 123 | ||
| GenBank mRNA | AK095020.1 | - | 1 - 2 | 122 | 123 | |
| BC035152.1 | - | 1 - 2 | 161 | 123 | ||
| DB487352.1 | - | 1 - 2 | 196 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000583.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16008532.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008532.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008532.hg.1 | TC16000583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008531.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008531.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008531.hg.1 | TC16000583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008530.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008530.hg.1 | TC16000583.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162480.hg.1 | TC1600008283.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600162478.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162478.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162478.hg.1 | TC1600008283.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600162479.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162479.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162479.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162479.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162479.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162479.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162479.hg.1 | TC1600008283.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162481.hg.1 | TC1600008283.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000398177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | ENST00000562507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008283.hg.1 | NR_034083.2 | Refseq |

VAC14-AS1
Genomic Map